Learn More
Medchemexpress LLC D-Fructose-d-1 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S111MG
Encompass_Preferred
D-Fructose-d-1 is the deuterium labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. This compound is intended for research use only, serving as a valuable tool in drug development and quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-004-19971