Learn More
Medchemexpress LLC D-Fructose-d4 | 478518-49-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S910MG
Encompass_Preferred
D-Fructose-d4 is the deuterium-labeled D-Fructose. D-Fructose is a naturally occurring monosaccharide found in many plants. It is for research use only and not for sale to patients.
  • Stable isotope
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Purity of 99.9%
Catalog No. 50-004-18079