Learn More
Medchemexpress LLC D-Fructose-d4 | 478518-49-9 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN7092S9
Encompass_Preferred
D-Fructose-d4 is the deuterium labeled D-Fructose, a naturally occurring monosaccharide found in many plants. Stable heavy isotopes have been incorporated as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Functions as a tracer.
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, or LC-MS).
  • Contains stable heavy isotopes for quantitation in drug development.
  • May influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-20428