Learn More
Medchemexpress LLC D-Fructose-d4 | 478518-49-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S95MG
Encompass_Preferred
D-Fructose-d4 is the deuterium labeled D-Fructose, a naturally occurring monosaccharide found in many plants.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-17645