Learn More
Medchemexpress LLC D-Fructose-d7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S1050MG
Encompass_Preferred
D-Fructose-d7 is the deuterium-labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. Intended for research use only, this compound is not for sale to patients. It can be utilized as a tracer and as an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, is used for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Deuterium-labeled form of D-Fructose
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-18965