Learn More
Medchemexpress LLC D-(+)-Fucose | 3615-37-0 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN5102100MG
Encompass_Preferred
D-(+)-Fucose is a nonmetabolizable analogue of L-arabinose. It prevents the growth of *Escherichia coli* B/r on a mineral salts medium plus L-arabinose by inhibiting the induction of the L-arabinose operon. D-fucose is also a potent inducer of beta-methylgalactoside permease (MGP).
  • Nonmetabolizable analogue of L-arabinose
  • Prevents growth of *Escherichia coli* B/r
  • Inhibits L-arabinose operon induction
  • Potent inducer of beta-methylgalactoside permease (MGP)
  • Can lead to transient repression of operon expression
  • Induces galactose transport
Catalog No. 50-004-20918