Learn More
Medchemexpress LLC D-(+)-Fucose | 3615-37-0 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN510250MG
Encompass_Preferred
D-(+)-Fucose is a nonmetabolizable analogue of L-arabinose. It prevents the growth of Escherichia coli B/r on a mineral salts medium plus L-arabinose by inhibiting the induction of the L-arabinose operon. D-fucose is also a potent inducer of beta-methylgalactoside permease (MGP). In vitro studies show that D-(+)-Fucose-resistant mutants produce an araC gene product with altered inducer specificity.
  • Nonmetabolizable analogue of L-arabinose.
  • Inhibits induction of the L-arabinose operon, preventing growth of *Escherichia coli* B/r.
  • Potent inducer of beta-methylgalactoside permease (MGP).
  • Alters inducer specificity of araC gene product in resistant mutants.
  • Causes transient repression of operon expression upon L-arabinose addition after D-(+)-Fucose induction.
  • Induces galactose transport in SB3586 cells in the presence of galactose or lactate.
Catalog No. 50-004-20953