Learn More
Cayman Chemical D-Fucose 500mg
SDP
Supplier:  Cayman Chemical 34343500 MG
Encompass_Preferred
A monosaccharide; inhibits induction of the L-arabinose operon; has been used to increase the yield and activity of recombinant proteins
Catalog No. 50-294-7079