Learn More
Medchemexpress LLC D-galacto-D-mannan (carob galactomannan) | 11078-30-1 | MFCD00146683 | 94.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW145481A5MG
Encompass_Preferred
D-Galacto-D-mannan (carob galactomannan) is a nonionic, linear polysaccharide derived from Ceratonia siliqua seeds. It is used in research as a thickener, stabilizer, co-gellant, and fat replacer, and is supplied as a powder for biochemical assay applications. CAS: 11078-30-1. Reported purity: 94%.
  • Nonionic linear polysaccharide derived from carob
  • Used as a thickener, stabilizer, co-gellant, and fat replacer
  • Reported purity 94%
  • Supplied as a powder suitable for biochemical assay use
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years)
Catalog No. 50-002-83002