Learn More
Medchemexpress LLC D-galacto-D-mannan from Ceratonia siliqua | 11078-30-1 | MFCD00146683 | 95.0% | 504.4 g/mol | C18H32O16 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW1454815MG
Encompass_Preferred
D-Galacto-D-mannan from Ceratonia siliqua is a plant cell wall polysaccharide (carob galactomannan) provided as a research-grade biochemical reagent with an indicated purity of 95.0%. The material is a heterogeneous polysaccharide used as a reference compound and reagent for analytical, biochemical, and immunological applications; molecular weight and polymer distribution may vary between batches.
  • Derived from Ceratonia siliqua (carob) plant material.
  • Supplied as a research-grade polysaccharide with approximately 95.0% purity.
  • Useful as a reference standard for analytical and compositional studies.
  • Suitable for biochemical and immunological applications.
  • Polysaccharide composition and molecular weight are variable; represents repeating galactomannan units.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-83003