Learn More
Medchemexpress LLC D-Galacto-D-mannan from Ceratonia siliqua | 11078-30-1 | MFCD00146683 | 95.0% | C18H32O16 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW14548125MG
Encompass_Preferred
D-Galacto-D-mannan from Ceratonia siliqua is a nonionic, linear plant cell-wall polysaccharide (carob galactomannan) used in research and formulation work as a thickener, stabilizer, co-gellant, and fat replacer. It is supplied as a powder of approximately 95.0% purity (CAS 11078-30-1) and has recommended powder storage of -20°C (3 years) or 4°C (2 years).
  • Nonionic, linear polysaccharide suitable for formulation studies.
  • Used as a thickener and stabilizer in biochemical assays.
  • Acts as a co-gellant and fat replacer in food and formulation research.
  • Provided as a powder with ~95.0% purity.
  • Stable in powder form when stored at -20°C for up to 3 years.
Catalog No. 50-002-83720