Learn More
Medchemexpress LLC α-D-Galactopyranose, 1,2:3,4-di-O-isopropylidene | 4064-06-6 | MFCD00063225 | 97.0% | 260.28 g·mol⁻¹ | C12H20O6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03664710G
Encompass_Preferred
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is a protected carbohydrate derivative used as a synthetic intermediate in organic and glycochemistry research. It temporarily masks the 1,2 and 3,4 hydroxyl groups of D-galactopyranose to enable selective transformations. Supplied as a colorless to light yellow liquid with supplier-stated purity around 97% and molecular weight 260.28 g·mol⁻1.
  • Protected hydroxyl groups enable selective reactions.
  • Suitable for glycosylation and carbohydrate synthesis.
  • High supplier-stated purity reduces need for additional purification.
  • Liquid form facilitates handling and dosing in small-scale synthesis.
  • Common building block for medicinal chemistry and glycochemistry.
Catalog No. 50-002-99740