Learn More
Medchemexpress LLC α-D-galactopyranosyl bromide, 2,3,4,6-tetraacetate | 3068-32-4 | MFCD00063686 | ≥97.0% | 411.20 g·mol⁻¹ | C14H19BrO9 | 25 G

Supplier: Medchemexpress LLC 25GHYW028813
Acetobromo-α-D-galactose (α-D-galactopyranosyl bromide, 2,3,4,6-tetraacetate) is a tetra-O-acetylated glycosyl bromide used as a brominating agent and glycosyl donor in carbohydrate chemistry. It is employed as an intermediate for the synthesis of glycosides and oligosaccharides and is provided as a powder with recommended cold storage.
- High purity (NMR ≥97.0%).
- Tetra-O-acetylated α-anomer suitable as a glycosyl donor.
- Powder form for convenient handling.
- Recommended storage: powder -20°C (stable for 3 years).
- In solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.