Learn More
Medchemexpress LLC D(+)-Galactosamine hydrochloride | 1772-03-8 | 99.9% | 215.63 | 25 MG
SDP
Catalog No. 5000402503
Encompass_Preferred
D(+)-Galactosamine hydrochloride is an established experimental toxin that primarily causes liver injury through the generation of free radicals and depletion of UTP nucleotides. Intoxication with D(+)-Galactosamine hydrochloride also induces renal dysfunction, with renal failure often associated with the end-stage of liver damage. The combination of Lipopolysaccharide/D(+)-Galactosamine is a known animal model for inducing acute liver injury, leading to fulminant hepatic failure.
  • Causes liver injury through generation of free radicals.
  • Leads to depletion of UTP nucleotides.
  • Induces renal dysfunction.
  • Useful in animal models for acute liver injury.
Catalog No. 50-004-02503 Supplier Medchemexpress LLC Supplier No. 25MGHY42682R
D(+)-Galactosamine hydrochloride is an established experimental toxin that primarily causes liver injury through the generation of free radicals and depletion of UTP nucleotides. Intoxication with D(+)-Galactosamine hydrochloride also induces renal dysfunction, with renal failure often associated with the end-stage of liver damage. The combination of Lipopolysaccharide/D(+)-Galactosamine is a known animal model for inducing acute liver injury, leading to fulminant hepatic failure.
  • Causes liver injury through generation of free radicals.
  • Leads to depletion of UTP nucleotides.
  • Induces renal dysfunction.
  • Useful in animal models for acute liver injury.