Learn More
Medchemexpress LLC D(+)-Galactosamine hydrochloride | 1772-03-8 | 99.9% | 215.63 | 500 MG
SDP
Catalog No. 5000404594
Encompass_Preferred
D(+)-Galactosamine hydrochloride is an established experimental toxin that primarily causes liver injury by generating free radicals and depleting UTP nucleotides. It also induces renal dysfunction, with renal failure often associated with the end-stage of liver damage. Lipopolysaccharide/D(+)-Galactosamine-induced acute liver injury is a known animal model of fulminant hepatic failure.
  • Induces apoptosis and necrosis in primary rat hepatocyte cultures.
  • Causes caspase-3 activation and DNA fragmentation.
  • Induces hepatotoxicity similar to viral hepatitis.
  • Suitable for research use only.
Catalog No. 50-004-04594 Supplier Medchemexpress LLC Supplier No. HY42682500MG
D(+)-Galactosamine hydrochloride is an established experimental toxin that primarily causes liver injury by generating free radicals and depleting UTP nucleotides. It also induces renal dysfunction, with renal failure often associated with the end-stage of liver damage. Lipopolysaccharide/D(+)-Galactosamine-induced acute liver injury is a known animal model of fulminant hepatic failure.
  • Induces apoptosis and necrosis in primary rat hepatocyte cultures.
  • Causes caspase-3 activation and DNA fragmentation.
  • Induces hepatotoxicity similar to viral hepatitis.
  • Suitable for research use only.