Learn More
Medchemexpress LLC D-Galactosamine pent 10g | 76375-60-5 | 389.35 g/mol | C16H23NO10 | 10 G
SDP
Supplier:  Medchemexpress LLC HY11727110G
Encompass_Preferred
D-Galactosamine pentaacetate is the penta-acetylated derivative of D-galactosamine used as a biochemical reagent and intermediate in carbohydrate and glycoconjugate synthesis. It is commonly used in organic synthesis and as a reference material for research applications.
  • Acetyl-protected amino sugar suitable for synthetic transformations.
  • Commonly used as a building block for glycoconjugate and carbohydrate chemistry.
  • Characterized by molecular weight 389.35 g/mol and formula C16H23NO10.
  • Typically supplied in a 10 g package with reported purity ~98%.
  • Provided with SDS documentation for safe handling and storage.
Catalog No. 50-002-27560