Learn More
Gold Biotechnology Inc D-Galactose 1 kg
SDP
Supplier:  Gold Biotechnology Inc G0101
Encompass
D-Galactose is a monosaccharide sugar that serves as an energy source and glycosylation component. It is a C-4 epimer of glucose and often used as a source of carbon in culture media. Galactose is a component of the disaccharide lactose and released upon hydrolysis by β-galactosidase enzymes. It is converted to glucose via the Leloir pathway or metabolized via an alternative pathway, such as the DeLey-Doudoroff pathway.
Catalog No. NC2596285