Learn More
Medchemexpress LLC D-glucaric acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 | C6H10O8 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW345885500MG
Encompass_Preferred
Saccharic acid (D-glucaric acid) is a research-grade small molecule used as a competitive inhibitor of β-glucuronidase and in biochemical assays. It is supplied as a solid with reported purity of 98.0%, CAS 87-73-0, formula C6H10O8, and molecular weight 210.14, typically packaged as a 500 mg vial for laboratory use.
  • High purity (98.0%).
  • Supplied as 500 mg solid.
  • Well-characterized; CAS 87-73-0.
  • Suitable as a β-glucuronidase inhibitor in biochemical assays.
  • Chemical formula C6H10O8; molecular weight 210.14.
Catalog No. 50-002-82205