Learn More
Medchemexpress LLC D-glucaric acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 g/mol | C6H10O8 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW3458851G
Encompass_Preferred
Saccharic acid (D-glucaric acid) is a small organic acid used in biochemical research as a competitive inhibitor of β-glucuronidase. Supplied as a white to off-white solid powder with high analytical purity, it is commonly used in enzymology, metabolism, and assay development.
  • High purity: 98.0% (NMR).
  • White to off-white solid powder for easy handling.
  • Competitive inhibitor of β-glucuronidase for enzymatic studies.
  • Molecular weight 210.14 g/mol, molecular formula C6H10O8.
  • Suitable for biochemical assays and metabolism research.
Catalog No. 50-002-80923