Learn More
Medchemexpress LLC D-glucaric acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 g/mol | C6H10O8 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW3458855G
Encompass_Preferred
Saccharic acid (D-glucaric acid) is a small-molecule competitive inhibitor of β-glucuronidase used in biochemical and metabolic research. It is supplied as a high-purity solid suitable for in vitro enzyme assays, inhibition studies, and investigations of metabolic pathways.
  • High purity (98.0%) for reproducible results.
  • Suitable for enzyme inhibition and metabolic assays.
  • Solid form, easily weighed and dissolved in appropriate solvents.
  • Well-characterized chemical identity (CAS 87-73-0) and molecular weight 210.14 g/mol.
  • Available in laboratory-scale pack sizes for experimental flexibility.
Catalog No. 50-002-80921