Learn More
Medchemexpress LLC D-Gluconic acid, calcium salt (2:1) | 299-28-5 | 98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB109210G
Encompass_Preferred
Gluconate Calcium is an orally active glucose derivative. It reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6), inhibits ERK phosphorylation, and possesses antioxidant and antiplatelet activation activities. It also exhibits antitumor activity against colorectal cancer and improves conditions such as osteoarthritis, intestinal damage, and acute lung injury.
  • Orally active glucose derivative
  • Reduces nitric oxide and inflammatory cytokines (IL-1β and IL-6)
  • Inhibits ERK phosphorylation
  • Has antioxidant and antiplatelet activation activities
  • Exhibits antitumor activity against colorectal cancer
  • Improves osteoarthritis, intestinal damage, and acute lung injury
Catalog No. 50-004-12462