Learn More
Medchemexpress LLC D-gluconic acid, gamma-lactone | 1198-69-2 | MFCD00080784 | >=95.0% | 178.14 g/mol | C6H10O6 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW047191
D-Glucono-1,4-lactone is a cyclic ester of D-gluconic acid used as a biochemical reagent in assays and research applications. It is supplied as a powder and should be stored refrigerated or frozen to maintain stability.
- white powder appearance
- molecular formula C6H10O6, molecular weight 178.14 g/mol
- purity (NMR) ≥95.0%
- CAS number 1198-69-2
- storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
- pack size: 250 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.