Learn More
Medchemexpress LLC Β-D-Glucopyranoside, 3-[6-(β-D-glucopyranosyloxy)-2-benzofuranyl]-5-hydroxyphenyl | 193483-95-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN35185MG
Encompass_Preferred
Mulberroside F is a bioactive constituent found in mulberry (Morus alba L.). It inhibits tyrosinase activity and melanin formation, and demonstrates superoxide scavenging activity to protect against auto-oxidation.
  • Inhibits tyrosinase activity
  • Reduces melanin formation
  • Exhibits superoxide scavenging activity
  • White to off-white solid
  • Purity of 99.37%
Catalog No. 50-004-18098