Learn More
Medchemexpress LLC α-D-Glucopyranoside, β-D-fructofuranosyl O-α-D-galactopyranosyl-(1→6)-, hydrate (1:5) | 17629-30-0 | 594.51 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN193810MM/1ML
Encompass_Preferred
D(+)-Raffinose pentahydrate is an orally active inhibitor of LecA and GtfC. It reduces cyclic diguanylate (c-di-GMP) by increasing the activity of a c-di-GMP specific phosphodiesterase. This compound interferes with GTF function, decreases IL-4 and IL-5 mRNA, and inhibits allergic airway eosinophilia.
  • Exhibits biofilm-inhibitory activity against Pseudomonas aeruginosa.
  • Exhibits biofilm-inhibitory activity against Streptococcus mutans.
  • Suppresses biofilm formation and reduces insoluble glucan production in Streptococcus mutans.
  • Downregulates gtfB and gtfC gene expression.
  • Alleviates allergen-induced airway eosinophilia in rats.
  • Reduces IL-4 and IL-5 mRNA levels in lung tissue.
  • Tends to lower alveolar macrophages, neutrophils, lymphocytes, and IgE levels.
Catalog No. 50-004-15398