Learn More
Medchemexpress LLC β-D-Glucopyranosiduronic, 5-hydroxy-8-methoxy-4-oxo-2-phenyl-4H-1-benzopyran-7-yl | 51059-44-0 | 460.39 | 1 ML

Supplier: Medchemexpress LLC HYN039910MM/1ML
Wogonoside, a flavonoid glycoside isolated from Huangqin, possesses anti-inflammatory effects and induces autophagy in breast cancer cells by regulating the MAPK-mTOR pathway.
- Isolated from Huangqin.
- Possesses anti-inflammatory effects.
- Induces autophagy in breast cancer cells.
- Regulates the MAPK-mTOR pathway.
- Purity of 99.91%.
- Appearance is light yellow to yellow solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.