Learn More
Medchemexpress LLC β-D-Glucopyranosiduronic acid, (3α,5α)-17-oxoandrostan-3-yl-2,2,4,4-d4 | 2687960-87-6 | C25H34D4O8 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY151027S1MG
Encompass_Preferred
Androsterone glucuronide-d4 is a deuterium-labeled form of Androsterone glucuronide. It is a white to off-white solid with a purity of 95.1% and is intended for research use only, not for medical applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Purity (HPLC) of 95.1%.
  • Store powder at -20°C for 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-74805