Learn More
Medchemexpress LLC β-D-Glucopyranosiduronic acid, 4-formyl-2-nitrophenyl, methyl ester, 2,3,4-triacetate | 148579-93-5 | 98.6% | 483.38 | C20H21NO13 | 500 MG

Supplier: Medchemexpress LLC HY131086500MG
Me-triacetyl-β-D-glucopyranosiduronic acid, 4-formyl-2-nitrophenyl, methyl ester, 2,3,4-triacetate is a cleavable glucuronide-based antibody-drug conjugate (ADC) linker used to connect antibody and payload during ADC synthesis.
- Cleavable glucuronide-based linker for ADC synthesis.
- High purity: 98.6% (LCMS).
- Cas number: 148579-93-5.
- Molecular weight: 483.38; formula C20H21NO13.
- Appearance: light yellow to yellow solid.
- Soluble in DMSO (200 mg/mL); in vivo formulations with solubility ≥ 2.5 mg/mL.
- Available in multiple packaging sizes, including 500 MG for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.