Learn More
Medchemexpress LLC β-D-glucopyranosiduronic acid methyl ester, 2,3,4-triacetate, Fmoc-protected phenylmethyl derivative | 894096-02-7 | MFCD34470435 | 98.8% | 748.73 g/mol | C38H40N2O14 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13108725MG
Encompass_Preferred
Me-triacetyl-β-D-glucopyranuronate-Ph-CH2OH-Fmoc is a glycosylated, Fmoc-protected cleavable linker used as a spacer in antibody-drug conjugate (ADC) synthesis to enable controlled release of payloads upon cleavage.
  • cleavable glycoside-based linker suitable for ADC conjugation
  • Fmoc-protected amine for orthogonal deprotection and conjugation workflows
  • high reported purity (98.8%) supporting research-grade use
  • available in small milligram quantities for synthetic and development work
  • molecular weight 748.73 g/mol and formula C38H40N2O14 for formulation and characterization
Catalog No. 50-002-02980