Learn More
Cayman Chemical D-GlucosamIn-6-sulfate 250mg
SDP
Supplier:  Cayman Chemical 23097250 MG
Encompass_Preferred
A glycosaminoglycan; activates the glmS ribozyme from B. subtilis (200 µM); has been used to form polyvalent dendrimer conjugates that inhibit angiogenesis, cell proliferation, and scar tissue formation
Catalog No. 50-296-4349