Learn More
Medchemexpress LLC D-Glucosamine 6-phosphate | 3616-42-0 | 259.15 | 50 MG

Supplier: Medchemexpress LLC HYN1184850MG
D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It can be used for the research of diabetes mellitus and appears as a white to off-white solid.
- Endogenous metabolite
- Can be used for the research of diabetes mellitus
- Available as a white to off-white solid
- Purity of 99.97%
- Soluble in water
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.