Learn More
Medchemexpress LLC D-Glucose-13C-1 | 84270-10-0 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S115MG
Encompass_Preferred
D-Glucose-13C-1 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a critical signaling molecule related to cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05397