Learn More
Medchemexpress LLC D-Glucose-13C | 101615-88-7 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S105MG
Encompass_Preferred
D-Glucose-13C is the 13C labeled form of D-Glucose. D-Glucose, also known as Glucose, is a monosaccharide and a crucial carbohydrate in biological systems. It functions as a carbohydrate sweetener and plays a vital role in general metabolism. Additionally, D-Glucose acts as a critical signaling molecule, linking cellular metabolic status with biotic and abiotic stress responses.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like carbon-13 are incorporated into drug molecules for quantitation during drug development.
  • Deuteration, a similar process, has been noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06505