Learn More
Medchemexpress LLC D-Glucose-13C-4 | 40762-22-9 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S16100MG
Encompass_Preferred
D-Glucose-13C-4 is a 13C labeled version of D-Glucose. D-Glucose, a monosaccharide, is a significant carbohydrate in biology. It functions as a carbohydrate sweetener, a vital component of general metabolism, and a crucial signaling molecule involved in cellular metabolic status and responses to biotic and abiotic stress. This product is intended solely for research purposes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as carbon, are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07320