Learn More
Medchemexpress LLC D-Glucose-13C-5 | 106032-62-6 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S17100MG
Encompass_Preferred
D-Glucose-13C-5 is the 13C labeled D-Glucose, a monosaccharide and important carbohydrate in biology. It acts as a carbohydrate sweetener, a vital component of general metabolism, and a critical signaling molecule involved in cellular metabolic status and biotic and abiotic stress responses.
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Important carbohydrate in biological processes
  • Acts as a carbohydrate sweetener
  • Critical component in general metabolism
  • Involved in cellular metabolic status signaling
  • Participates in biotic and abiotic stress responses
Catalog No. 50-004-07268