Learn More
Medchemexpress LLC D-Glucose-13C-5 | 106032-62-6 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S175MG
Encompass_Preferred
D-Glucose-13C-5 is the 13C labeled D-Glucose. It is a monosaccharide, an important carbohydrate in biology, a carbohydrate sweetener, and a critical component of general metabolism. It also serves as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06503