Learn More
Medchemexpress LLC D-Glucose-13C6 | 110187-42-3 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0389A5G
Encompass_Preferred
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose (HY-B0389). It can be used as a metabolic tracer to track glucose-related synthetic catabolism, or as a synthesis ingredient, minimal media reagent, and internal standard. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07123