Learn More
Sigma Aldrich Fine Chemicals Biosciences D-(+)-Glucose, 50-99-7, MFCD00063774, 1 kg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G70211KG
Encompass
Empirical Formula (Hill Notation): C6H12O6, Molecular Weight: 180.16, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%, Synonym: Dextrose. D-(+)-Glucose has been used for culturing of mammalian cells. It has been used to study interactions between glucose and alanine and their influence on pentose phosphate pathway activity. It has been used to perform glucose tolerance test in mice.
Catalog No. NC0619478