Learn More
Medchemexpress LLC D-glucose-d1 | 56570-89-9 | MFCD35069281 | 99.9% | 181.16 g/mol | C6H11DO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S110MG
Encompass_Preferred
D-glucose-d1 is a monodeuterated form of D-glucose (one hydrogen replaced by deuterium) used as an isotopic tracer in biochemical and metabolic studies. It serves as a labeled substrate for tracing glucose uptake and metabolic flux, and as an analytical standard in mass spectrometry and NMR applications.
  • Monodeuterated (d1) isotopic tracer for metabolic studies.
  • High chemical purity (99.9%).
  • Molecular weight 181.16 g/mol; formula C6H11DO6.
  • Suitable for analytical standards, mass spectrometry, and NMR experiments.
  • Available in small research pack sizes (e.g., 10 MG) for laboratory studies.
Catalog No. 50-002-97116