Learn More
Medchemexpress LLC D-Glucose-d12-1 | 89798-27-6 | 99.4% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0389S2
Encompass_Preferred
D-Glucose-d12-1 is the deuterium labeled D-Glucose. D-Glucose, a monosaccharide, is an important carbohydrate in biology, serving as a critical component of general metabolism and a signaling molecule related to cellular metabolic status and stress response. Stable heavy isotopes of hydrogen, carbon, and other elements, such as deuterium, are incorporated into drug molecules for quantitation during drug development due to their potential to affect pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-50559