Learn More
Medchemexpress LLC D-Glucose-d2 | 18991-62-3 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S5100MG
Encompass_Preferred
D-Glucose-d2 is the deuterium labeled D-Glucose. D-Glucose, a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a signaling molecule in relation to cellular metabolic status and biotic/abiotic stress response.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07146