Learn More
Medchemexpress LLC D-Glucose-d7 | 66034-51-3 | 99.4% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S650MG
Encompass_Preferred
D-Glucose-d7 is the deuterium labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07347