Learn More
Alkali Scientific D (+) Glucose [Dextrose Anhydrous], USP Grade, 500 G
SDP
Supplier:  Alkali Scientific COS004CP
Encompass_Preferred
D (+) Glucose Anhydrous (USP Grade), 500 g, is a high-purity carbohydrate reagent commonly used in laboratory research, pharmaceutical applications, and biotechnology. Glucose is a critical metabolic substrate for cellular energy production and is essential in the study of cellular respiration, fermentation, and enzyme kinetics. The anhydrous form of glucose is preferred in situations where water content is unwanted or needs to be precisely controlled, making it ideal for dry chemical processes, media preparation, and biochemical assays. This 500 g quantity offers a convenient size for smaller-scale laboratory experiments and applications.
Catalog No. 50-006-60768