Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucuronic acid sodium monohydrate-13C6 | 207300-70-7 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN6612S5MG

Encompass_Preferred

D-Glucuronic acid sodium monohydrate-13C6 is the 13C-labeled version of D-Glucuronic acid sodium salt monohydrate. D-Glucuronic acid sodium salt monohydrate is an endogenous metabolite.

  • Product type: Stable isotope
  • Purity: 99.9%
  • Molecular weight: 240.09
  • Formula: 13C6H11NaO8
  • Appearance: Solid
  • Color: White to off-white
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements are used as tracers for drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • Storage (powder): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • For research use only
  • Documentation: Data Sheet, COA, SDS, handling instructions available

Catalog No. 50-004-17877


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.