Learn More
Medchemexpress LLC D-glutamic acid | 6893-26-1 | MFCD00063112 | 147.13 g/mol | C5H9NO4 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10080525G
Encompass_Preferred
D-Glutamic acid is the D-enantiomer of glutamic acid supplied as a white powder for research and analytical use. It has the chemical formula C5H9NO4, molecular weight 147.13 g/mol, and CAS number 6893-26-1. The compound is commonly used as a reference standard and reagent in pharmaceutical, biochemical, and food research and can be dissolved in aqueous buffers with sonication and pH adjustment.
  • High chemical identity as the D-enantiomer.
  • Well characterized: CAS 6893-26-1 and molecular weight 147.13 g/mol.
  • Powder form suitable for weighing and formulation of stock solutions.
  • Water solubility with recommended sonication and pH adjustment for stock preparation.
  • Stable when stored at recommended temperatures for extended shelf life.
Catalog No. 50-002-12195