Learn More
Medchemexpress LLC D-glutamic acid-d5 | 14341-88-9 | MFCD08704921 | 99.9% | 152.16 g·mol⁻¹ | C5H4D5NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100805S10MG
Encompass_Preferred
D-Glutamic acid-d5 is the deuterium-labeled (d5) isotopologue of D-glutamic acid supplied as a high-purity analytical standard. It is the D-enantiomer of glutamic acid and is intended for use as an internal standard in quantitative mass spectrometry and other analytical workflows for pharmaceutical and food research.
  • Deuterium-labeled (d5) isotopologue for clear mass shift in MS.
  • High purity (~99.9%) suitable for quantitative analysis.
  • Suitable as internal standard for LC-MS and GC-MS methods.
  • Useful for method development, validation, and trace quantification.
  • Stable solid material convenient for storage and handling.
Catalog No. 50-002-13421