Learn More
Medchemexpress LLC D-glutamine | 5959-95-5 | MFCD00065607 | 98.0% | 146.14 g/mol | C5H10N2O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1005871G
Encompass_Preferred
D-Glutamine is the D-stereoisomer of the amino acid glutamine provided for laboratory and analytical research. It is used to probe amino acid metabolism, participate in studies of neurotransmitter cycling, and model effects on epithelial barrier function. Not for human or clinical use.
  • Cell-permeable D-stereoisomer of glutamine.
  • Useful for studies of the glutamate/GABA-glutamine cycle and neuronal metabolism.
  • Used to evaluate protection against acetaldehyde-induced barrier disruption in Caco-2 models.
  • Analytical standard quality with reported purity 98.0%.
  • Molecular formula C5H10N2O3; molecular weight 146.14 g/mol.
  • Intended for research use only; not for sale to patients.
Catalog No. 50-002-14770