Learn More
Medchemexpress LLC D-glutamine | 5959-95-5 | MFCD00065607 | >98.0% | 146.14 g/mol | C5H10N2O3 | 10 G

Supplier: Medchemexpress LLC HY10058710G
D-Glutamine is the D-stereoisomer of the amino acid glutamine, provided as a research-grade reagent and analytical standard. It is cell-permeable and commonly used in metabolic and cellular studies.
- Purity >98%.
- CAS number 5959-95-5.
- Molecular weight 146.14 g/mol.
- Solid powder form suitable for cell-permeability studies.
- Available in multiple pack sizes, including 10 g.
- Used in metabolic and cellular research and as an analytical standard.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.