Learn More
Medchemexpress LLC D-Glutamine | 5959-95-5 | MFCD00065607 | 98.0% | 146.14 g/mol | C5H10N2O3 | 5 G

Supplier: Medchemexpress LLC HY1005875G
D-Glutamine is the D-stereoisomer of the amino acid glutamine supplied as an analytical standard for research and analytical applications. It is cell-permeable and is provided in solid and solution formats suitable for stereospecific metabolic studies, analytical reference, and cell-based experiments.
- Purity 98.0% by HPLC.
- Molecular formula C5H10N2O3.
- Molecular weight 146.14 g/mol.
- CAS number 5959-95-5.
- Available in multiple package sizes and solution formats.
- Intended for research and analytical use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.