Learn More
Medchemexpress LLC D-I03 | 688342-78-1 | 99.7% | 428.64 g/mol | C23H36N6S | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12469110MM 1ML
Encompass_Preferred
D-I03 is a small-molecule RAD52 inhibitor used in research to block RAD52-dependent single-strand annealing and D-loop formation. It is supplied as an off-white to light-yellow solid and as a ready-to-use 10 mM solution in DMSO, with high purity and recommended storage conditions for powder and solutions.
  • Selective inhibition of RAD52-mediated ssDNA annealing.
  • Demonstrated biochemical activity with reported IC50 and Kd values.
  • Available as solid and as a 10 mM solution in DMSO for workflow flexibility.
  • High purity suitable for research applications.
  • Stable under recommended storage conditions for long-term use.
  • Molecular formula C23H36N6S; molecular weight 428.64 g/mol.
Catalog No. 50-002-37947