Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-iditol | 25878-23-3 | MFCD09951922 | 98.0% | 182.17 | C6H14O6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW1455845MG

Encompass_Preferred

D-Iditol is a fungal metabolite and sugar alcohol used in biochemical and metabolic research. It accumulates in galactokinase deficiency and has reported potential antitumour activity. The compound is supplied as a solid for laboratory studies and analytical use.

  • Purity 98.0%.
  • Molecular formula C6H14O6; molecular weight 182.17.
  • Available in small research sizes including 5 mg.
  • Storage 4°C, sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Suitable for metabolic studies and exploratory antitumour research.

Catalog No. 50-002-83842


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.